Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029711.1 | NP_083987.1 | 5 | 696 | ||
GenBank mRNA | AK008777.1 | - | 1 | 142 | ||
AK037662.1 | - | 1 | 1385 | |||
AK083263.1 | - | 5 | 506 | |||
AK151513.1 | - | 5 | 670 | |||
AK152356.1 | - | 6 | 521 | |||
AK170061.1 | - | 6 | 495 | |||
BC099504.1 | - | 4 | 348 | |||
BC115750.1 | - | 5 | 494 | 40051600 |