Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029934.3 | NP_084210.2 | 7 | 1805 | ||
GenBank mRNA | AB055410.1 | - | 7 | 1803 | ||
AK019981.1 | - | 7 | 1216 | |||
AK038998.1 | - | 6 | 2920 | |||
AK154606.1 | - | 7 | 1804 | |||
EU016380.1 | - | 6 | 1017 |