Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207633.2 | NP_997516.1 | 3 | 698 | ||
GenBank mRNA | AK020780.1 | - | 3 | 272 | ||
AK030473.1 | - | 3 | 700 | |||
AK033612.1 | - | 1 | 277 | |||
AK045453.1 | - | 3 | 410 | |||
AK053562.1 | - | 3 | 699 | |||
AK079683.1 | - | 3 | 310 | |||
AK143152.1 | - | 3 | 268 | |||
AK160251.1 | - | 4 | 326 | |||
BC048712.1 | - | 3 | 333 | 6328657 |