Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030052.3 | NP_084328.1 | 3 | 256 | ||
GenBank mRNA | AK019665.1 | - | 3 | 257 | ||
AK076833.1 | - | 3 | 257 | |||
BC059709.1 | - | 3 | 251 | 6741029 | ||
BY099104.1 | - | 3 | 256 |