Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289780.1 | NP_001276709.1 | 15 | 1913 | ||
NM_001289781.1 | NP_001276710.1 | 14 | 2246 | |||
NM_178403.5 | NP_848490.2 | 14 | 1895 | |||
GenBank mRNA | AK021205.1 | - | 254 | |||
AK142271.1 | - | 14 | 1864 | |||
AK145380.1 | - | 14 | 2246 | |||
AK160849.1 | - | 14 | 1847 | |||
AK173290.1 | - | 14 | 1835 | |||
AK180496.1 | - | 2 | 113 | |||
BC008544.1 | - | 14 | 1849 | 3497634 | ||
BC010600.1 | - | 6 | 630 | |||
BC138533.1 | - | 14 | 1737 | 8861553 | ||
BC145316.1 | - | 15 | 1755 | 9053871 |