Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cptp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024472.4 | NP_077792.2 | 3 | 840 | ||
GenBank mRNA | AK040664.1 | - | 3 | 841 | ||
AK089229.1 | - | 3 | 590 | |||
BC002216.1 | - | 2 | 532 | 3490501 |