Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271725.1 | NP_001258654.1 | 12 | 2304 | ||
NM_001271726.1 | NP_001258655.1 | 12 | 2340 | |||
NM_001271727.1 | NP_001258656.1 | 12 | 2609 | |||
NM_001271728.1 | NP_001258657.1 | 12 | 2407 | |||
NM_030706.3 | NP_109631.2 | 12 | 2403 | |||
GenBank mRNA | AB043550.1 | - | 12 | 2387 | ||
AF220017.1 | - | 12 | 2252 | |||
AK042752.1 | - | 4 | 396 | |||
AK045410.1 | - | 12 | 2400 | |||
AK079415.1 | - | 12 | 2554 | |||
AK141419.1 | - | 12 | 2250 | |||
AK147275.1 | - | 12 | 2517 | |||
AK147395.1 | - | 12 | 2398 | |||
AK164350.1 | - | 11 | 2106 | |||
AK220343.1 | - | 11 | 3846 | |||
BC058668.1 | - | 11 | 3215 | |||
BC058961.1 | - | 12 | 2407 | 6826143 |