Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148941.2 | NP_683743.2 | 6 | 1123 | ||
GenBank mRNA | AF277093.1 | - | 6 | 919 | ||
AK029065.1 | - | 6 | 984 | |||
AK030666.1 | - | 6 | 1072 | |||
BC037030.1 | - | 6 | 1066 | 4503493 |