Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033565.2 | NP_291043.1 | 16 | 3213 | ||
GenBank mRNA | AF190449.1 | - | 16 | 3155 | ||
AK033163.1 | - | 6 | 1033 | |||
AK053034.1 | - | 4 | 243 | |||
BC138999.1 | - | 16 | 2886 | 8862021 | ||
BC145490.1 | - | 15 | 2799 | 9054053 |