Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45gip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183358.4 | NP_899202.3 | 1 | 298 | ||
GenBank mRNA | AK002738.1 | - | 1 | 304 | ||
AK003314.1 | - | 1 | 299 | |||
AK009723.1 | - | 1 | 297 | |||
AK019570.1 | - | 1 | 299 | |||
AK028227.1 | - | 1 | 299 | |||
BC022592.1 | - | 1 | 183 | |||
BC061069.1 | - | 1 | 298 | 6740575 |