siRNA Details
Gene Symbol: Aida | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181732.4 | NP_859421.1 | 10 | 1295 | ||
GenBank mRNA | AK027971.1 | - | 4 | 346 | ||
AK081494.1 | - | 10 | 1296 | |||
AK085039.1 | - | 8 | 789 | |||
AK143716.1 | - | 10 | 835 | |||
AK149788.1 | - | 10 | 1047 | |||
AK150639.1 | - | 10 | 983 | |||
AK150702.1 | - | 10 | 1044 | |||
AK151783.1 | - | 10 | 985 | |||
AK152961.1 | - | 10 | 986 | |||
AK153448.1 | - | 10 | 983 | |||
AK158103.1 | - | 10 | 929 | |||
AK159318.1 | - | 10 | 1307 | |||
AK167520.1 | - | 10 | 1012 | |||
AK172451.1 | - | 10 | 988 | |||
BC057183.1 | - | 8 | 704 | 5065689 | ||
BC066829.1 | - | 10 | 834 | |||
BC086763.1 | - | 10 | 1000 | 5707830 |

More Information
Other Species Targeted

