Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133357.3 | NP_579935.1 | 2, 3 | 792 | ||
GenBank mRNA | AF343088.1 | - | 1, 2 | 210 | ||
AK028972.1 | - | 2, 3 | 793 | |||
AK037233.1 | - | 2, 3 | 793 | |||
AK076367.1 | - | 2, 3 | 793 | |||
BC137935.1 | - | 2, 3 | 793 | 8860954 | ||
BC137936.1 | - | 2, 3 | 793 | 8860955 |