Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052835.3 | NP_443067.1 | 5 | 366 | ||
GenBank mRNA | AK011022.1 | - | 5 | 326 | ||
AK012561.1 | - | 4 | 323 | |||
AK014054.1 | - | 5 | 328 | |||
AK088777.1 | - | 5 | 327 | |||
AK144831.1 | - | 5 | 326 | |||
AK146217.1 | - | 5 | 327 | |||
AK151416.1 | - | 5 | 326 | |||
AK151655.1 | - | 5 | 326 | |||
AK151736.1 | - | 5 | 325 | |||
AK152123.1 | - | 5 | 326 | |||
AK152854.1 | - | 5 | 326 | |||
AK153345.1 | - | 5 | 326 | |||
AK160618.1 | - | 5 | 326 | |||
AK166610.1 | - | 5 | 325 | |||
AK167239.1 | - | 4 | 362 | |||
AK167966.1 | - | 5 | 326 | |||
AK168250.1 | - | 5 | 325 | |||
AK168854.1 | - | 5 | 326 | |||
AK187585.1 | - | 3 | 257 | |||
AK187587.1 | - | 3 | 219 | |||
AK210081.1 | - | 3 | 215 | |||
BC006068.1 | - | 4 | 285 | |||
BC024901.1 | - | 4 | 287 | 4224016 | ||
BC048872.1 | - | 4 | 290 | 5052784 | ||
BC082293.1 | - | 4 | 287 | 6486491 | ||
BC083327.1 | - | 4 | 287 | 5684015 | ||
BC086917.1 | - | 5 | 325 | 6768088 | ||
BC092383.1 | - | 4 | 311 | 5710462 | ||
BC098204.1 | - | 4 | 311 | 6410755 | ||
BC099437.1 | - | 4 | 290 | 30843551 | ||
M93980.1 | - | 5 | 325 | |||
U89405.1 | - | 2 | 195 | |||
X75312.1 | - | 4 | 318 |