ARL™ PERFORM'X Sequential X-Ray Fluorescence Spectrometer
ARL™ PERFORM'X Sequential X-Ray Fluorescence Spectrometer
ARL™ PERFORM'X Sequential X-Ray Fluorescence Spectrometer
ARL™ PERFORM'X Sequential X-Ray Fluorescence Spectrometer
Thermo Scientific™
ARL™ PERFORM'X Sequential X-Ray Fluorescence Spectrometer
Solve analytical challenges in a wide range of applications with X-ray fluorescence with sample surface mapping and small spot analysis.
Have Questions?
Catalog NumberType
IQLAAHGABMFALXMAZESimultaneous-Sequential XRF Spectrometer
Catalog number IQLAAHGABMFALXMAZE
Price (USD)
-
Type:
Simultaneous-Sequential XRF Spectrometer
Solve analytical challenges in a wide range of applications with one comprehensive tool. The Thermo Scientific™ ARL™ PERFORM'X Sequential X-Ray Fluorescence Spectrometer for advanced materials characterization integrates bulk elemental analysis with mapping and small spot analysis to create a solution that evaluates up to 90 elements in nearly any solid or liquid sample. Industries as diverse as metallurgy, petroleum, polymers, mining, glass, cement and refractories as well as labs dealing with geochemistry, materials science, environmental research and forensics will benefit from its performance and versatility.
With the ARL PERFORM'X spectrometer, analysis of materials in varied sizes, coatings, layers, heterogeneities and inclusions or requiring wide elemental coverage, wide concentration ranges and varied samples matrices is achieved with desired precision and limits of detection.
Advanced Technology
  • Digitally mastered gearless goniometer based on Moiré fringe electro-optical technology, for precise and robust angular positioning of all its components.
  • Powerful OXSAS software and standard-less Thermo Scientific™ QuantAS™ and Thermo Scientific™ UniQuant™ Analysis Packages provide simple and rapid analysis of totally unknown samples with no standards required.
  • Thermo Scientific™ PetroilQuant™ Pre-calibration program, capable of quantifying up to 30 elements in a variety of fuels, light and heavy distillates and lubricants both quickly and cost-effectively.
  • UCCO technology: Ultra Closely Coupled Optics for highest performance from boron to uranium.
  • IQ/OQ & Remote Diagnostics for easy installation and serviceability.
Main Features
  • Large library of factory calibrations covering many metals and non-metals applications.
  • Material composition mapping and quantification capability.
  • Small spot analysis for contamination or defect identification and quantification.
  • Industry-leading liquid sample recognition and leak prevention design.
  • Wide dynamic range (from sub ppm to 100%) and excellent precision.
  • Integrated comprehensive sample changer allows large numbers of different samples to be treated fully automatically in mixed batches.
  • Faster analysis cycle time for high throughput lab applications using XY-loader for fully automated sample management.
  • Easy, non-destructive sample preparation.
ARL PERFORM'X 1500W: Cost effective low-power system, no external water cooling required.
ARL PERFORM'X 2500W: High performance mid-power system, no external water cooling required.
ARL PERFORM'X 4200W: Highest power system for the most demanding applications.
Recommended for
  • Geological and environmental applications.
  • Petroleum products, polymers and chemicals.
  • Materials science and forensics.
  • Major, minor and trace elemental analysis in raw materials, glasses, ceramics and refractories.
  • Specialty alloys, metals and ores.
  • Mining, minerals and cement.
Specifications
TypeSimultaneous-Sequential XRF Spectrometer
Unit SizeEach
Frequently asked questions (FAQs)
What advantages does WDXRF provide for complex sample matrices?
WDXRF provides higher spectral resolution than EDXRF, helping reduce peak overlaps and improve elemental separation in complex sample matrices. This can support more accurate quantification when analyzing samples containing multiple elements with overlapping spectral characteristics.
What factors should I consider when selecting a WDXRF spectrometer?
When selecting a WDXRF spectrometer, laboratories typically evaluate sample types, required detection limits, elemental coverage, throughput requirements, automation needs, software capabilities, regulatory requirements, and long-term analytical objectives. Matching instrument capabilities to application requirements can help optimize laboratory performance and workflow efficiency.
Can the ARL PERFORM'X support both routine quality control and advanced research workflows?
Yes. The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer is designed to support routine production testing as well as advanced research applications. Its combination of broad elemental coverage, flexible analytical capabilities, and quantitative performance allows laboratories to use a single system across multiple workflows.
What capabilities make the ARL PERFORM'X suitable for trace elemental analysis?
The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer combines high spectral resolution, excellent precision, broad dynamic range, and low detection limits to support demanding trace elemental analysis applications. These capabilities make it suitable for quality control, regulatory compliance, raw material characterization, environmental analysis, and advanced research workflows.
Which laboratories are best suited for the ARL PERFORM'X WDXRF Spectrometer?
The Thermo Scientific ARL PERFORM'X Sequential X-Ray Fluorescence (WDXRF) Spectrometer is well suited for laboratories that require high-precision elemental analysis, including mining, metals, cement, petrochemical, environmental, geological, materials science, and industrial quality control laboratories.