Explore how PGNAA and PFTNA work and the key features of hardware and software that support its industrial use
Welcome to our Learning Center for PGNAA, an analytical technique used by the processing industries, notably the coal, cement, minerals processing and steel sectors, for non-contact, non-destructive online measurement of the elemental composition of bulk raw materials. 
PGNAA is the acronym for Prompt Gamma Neutron Activation Analysis. It’s a technique that requires a supply of neutrons which can come from either a radioactive source, typically Cf-252, or a neutron generator via Pulsed Fast Thermo Neutron Activation (PFTNA). We pioneered PGNAA and are highly experienced in its  successful application in demanding commodity industries.
Here’s where we share our expertise and knowledge. Find the valuable information you need to accelerate and optimize the application of both PGNAA and PFTNA by browsing through the educational assets below.
Quick links
Learn more about technologies being utilized, resources about our offerings, and frequently asked questions from peers in your field.
PGNAA technology
PGNAA FAQs
PGNAA resources
Featured assets
See here for access to several helpful documents and videos, visit the resources tab above for a larger selection.
eBook: Guide to PGNAA and PFTNA technology for non-scientists
All the basics to get you started, from an introduction to the technique to an overview of how the technology  can help miners, mineral and coal processors, and cement and steel producers.
Infographic: PGNAA detection limit guidelines
See just how many elements PGNAA can measure with ease and accuracy, and examine the detection limits you can expect.
Video: CB Omni Agile Online Elemental Analyzer
Take a closer look at PGNAA technology, at how it integrates with process plant and the features that determine performance.
Webcast: Technology considerations for bulk ore sorting
Learn about the unique benefits of PGNAA for realizing the economic and environmental potential of bulk ore sorting.
Brochure: Cost-efficient strategies for sustainable cement production
Find out how PGNAA is helping cement manufacturers to operate smarter and more economically as the drive to reduce environmental impact intensifies.
Success story: Building green mines to deliver economic and social benefits.
Read how Chinese companies are using PGNAA technology to digitize limestone mining and reap substantial economic and environmental gain.
From rock to results: Real-time insights with PGNAA
Join Kevin Gordon and Garry Noble as they uncover how PGNAA delivers real-time elemental insights that transform raw material variability into actionable process intelligence.
40 years of PGNAA technology
For 40 years, Thermo Fisher Scientific has advanced PGNAA technology to provide continuous, real-time insight into material composition, evolving from early chute-based systems to intelligent analyzers that enable data-driven process control.
For Research Use Only. Not for use in diagnostic procedures.