    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • CCL21 Proteins
          Gibco
          Human CCL21 (6Ckine) Recombinant Protein, PeproTech®
          3 References
          View all (4) CCL21 proteins
          Tech Support
          Cite Human CCL21 (6Ckine) Recombinant Protein, PeproTech®
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Testing Data (1)
          Human CCL21 (6Ckine) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Human CCL21 (6Ckine) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Human CCL21 (6Ckine) Protein (300-35A-1MG) in Bioactivity
          Bioassay analysis of Human CCL21 (6Ckine) Recombinant Protein, PeproTech® (Product # 300-35A-1MG). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human CCL21 (6Ckine) Protein in Bioactivity (Bioactivity)
          Product Details
          300-35A-1MG
          Applications
          Tested Dilution
          Publications
          Control (Ctrl)
          Assay-dependent
          -
          In vitro Assay (In vitro)
          -
          View 2 publications 2 publications
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Bioactivity (Bioactivity)
          Assay-dependent
          -
          Product Specifications
          Species
          Human
          Published species
          Human, Mouse
          Expression System
          E. coli
          Amino acid sequence
          SDGGAQDCCL KYSQRKIPAK VVRSYRKQEP SLGCSIPAIL FLPRKRSQAE LCADPKELWV QQLMQHLDKT PSPQKPAQGC RKDRGASKTG KKGKGSKGCK RTERSQTPKG P
          Molecular weight
          12.2 kDa
          Class
          Recombinant
          Type
          Protein
          Purity
          ≥ 98% by SDS-PAGE gel and HPLC analyses.
          Endotoxin concentration
          <1 EU/µg
          Activity
          Determined by its ability to chemoattract human activated T cells cultured in the presence of IL-2.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Amount
          1 mg
          Purification
          purified
          Contains
          no preservative
          Storage conditions
          -20°C
          Shipping conditions
          Ambient
          Product Specific Information
          Recombinant Human Exodus-2 (CCL21) is a 12.2 kDa protein containing 111 amino acid residues. *Human Exodus-2 (Catalog Number 300-35A) has replaced Human Exodus-2 (Catalog Number 300-35).
          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
          Target Information
          This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
          Bioinformatics
          Protein Aliases: 6 Ckine; 6Ckine; CC chemokine; beta chemokine exodus-2; CCL21; chemokine (C-C motif) ligand 21; Efficient Chemoattractant for Lymphocytes; secondary lymphoid tissue chemokine; small inducible cytokine subfamily A (Cys-Cys), member 21; unnamed protein product
          View more View less
          Gene Aliases: 6Ckine; CKb9; ECL; SCYA21; SLC; TCA4
          View more View less
          UniProt ID: (Human) O00585
          View more View less
          Entrez Gene ID: (Human) 6366
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch