Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55350. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The yeast heterotetrameric GINS complex is made up of Sld5, Psf1 (GINS1), Psf2 (GINS2), and Psf3 (GINS3). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SLD5 homolog; unnamed protein product
Gene Aliases: SLD5
UniProt ID: (Human) Q9BRT9
Entrez Gene ID: (Human) 84296
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support