Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62365. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Involved in the lipid remodeling steps of GPI-anchor maturation. Lipid remodeling steps consist in the generation of 2 saturated fatty chains at the sn-2 position of GPI-anchor proteins (GPI-AP). Has phospholipase A2 activity that removes an acyl-chain at the sn-2 position of GPI-anchors during the remodeling of GPI. Required for the shedding of the GPI-AP TDGF1, but not CFC1, at the cell surface. Shedding of TDGF1 modulates Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells (PubMed:27881714). Also indirectly involved in the translocation of RAC1 from the cytosol to the plasma membrane by maintaining the steady state amount of CAV1-enriched plasma membrane subdomains, stabilizing RAC1 at the plasma membrane (PubMed:27835684). In contrast to myomaker (TMEM8C), has no fusogenic activity (PubMed:26858401). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: five-span transmembrane protein M83; GPI processing phospholipase A2; post-glycosylphosphatidylinositol attachment to proteins 6; transmembrane protein 6; transmembrane protein 8 (five membrane-spanning domains); transmembrane protein 8A; unnamed protein product
Gene Aliases: GPI-PLA2; M83; TMEM6; TMEM8; TMEM8A
UniProt ID: (Human) Q9HCN3
Entrez Gene ID: (Human) 58986
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support