Hamburger Menu Button
Thermo Fisher Scientific Logo
로그인
회원이 아니신가요? 계정 생성하기
  • 모든 제품 주문
    • 항체
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR 분석
    • 피펫 및 피펫 팁
    • 실험실용 원심분리기
    • 초저온 냉동고
    • 분광학
    • 비이커
    • PCR 장비 및 용품
    • 제품 카테고리 전체보기
  • 응용 분야 및 기법
    • 세포 분석
    • 실험실 장비
    • Real-Time PCR
    • PCR
    • 크로마토그래피
    • 세포 배양 및 트랜스펙션
    • DNA/RNA 추출과 분석
    • 단백질 생물학
    • 유세포분석
    • 화학 제품
    • 응용 분야 및 기법 전체보기
  • 서비스
    • 맞춤형 서비스
    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 신제품
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo

Search

전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          • Proteins & Peptides ›
          • GM-CSF Proteins

          Gibco

          Mouse GM-CSF Recombinant Protein

          View all (16) GM-CSF proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Mouse GM-CSF Recombinant Protein

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          PMC2015

          Applications
          Tested Dilution
          Publications

          Bioactivity (Bioactivity)

          Assay-dependent
          -
          Product Specifications

          Species

          Mouse

          Expression System

          E. coli

          Molecular weight

          14 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          >95% by SDS-PAGE

          Endotoxin concentration

          <0.1 ng/µg

          Activity

          ED50 < 0.01 ng/mL; determined by the dose-dependent proliferation of MC/9 cells.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          10 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Carrier-Free

          Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Apportion the reconstituted protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in low endotoxin medium or a buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA.

          Storage: Store the lyophilized protein at 2-8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze-thaw cycles.

          Target Information

          The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: colony-stimulating factor; granulocyte macrophage colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte/macrophage colony-stimulating factor; M-GM-CSF; MGC131935; MGC138897; MGC151255; MGC151257; put. GM-CSF; unnamed protein product

          View more View less

          Gene Aliases: CSF; Csfgm; Gm-CSf; GMCSF; MGI-IGM

          View more View less

          UniProt ID: (Mouse) P01587

          View more View less

          Entrez Gene ID: (Mouse) 12981

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          About Thermo Fisher Plus Icon Minus Icon
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)

           

          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)

           

          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-l699q:80/100.66.79.160:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline