Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 1000.0 | 9.009009 |
| Magnesium Chloride (MgCl2-6H2O) | 203.0 | 1000.0 | 4.9261084 |
| Potassium Chloride (KCl) | 75.0 | 2000.0 | 26.666666 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 2000.0 | 14.705882 |
| Sodium Chloride (NaCl) | 58.0 | 80000.0 | 1379.3103 |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 21600.0 | 80.597015 |
1. Dulbecco, R. and Vogt, M., (1954) Plaque formation and isolation of pure lines with Poliomyelitis viruses. J. Exp. Med., 98:167.
14040 - DPBS, calcium, magnesium
Cat No:
14040083
,
14040091
,
14040117
,
14040133
,
14040141
,
14040166
,
14040174
,
14040182
,
14040216
,
14040224
,
14040224
21300 - DPBS, powder, calcium, magnesium
Cat No:
21300025
,
21300058
,
21300058
,
21300074