Schneider's Drosophila Medium, 500 mL - FAQs
View additional product information for Schneider's Drosophila Medium - FAQs (21720024, 21720001)
1 product FAQs found
Are D. Mel-2 cells the same as Schneider S2 cells?
D. Mel-2 (Drosophila melanogaster-2) cells are basically Schneider S2 cells that have been adapted to growth in serum-free media.