Search
Search

SMO1-2| Gene Symbol: SMO1-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118404.5 | NP_567670.1 | 3 - 4 | 1034 | 126 | |
| GenBank mRNA | AK229338.1 | - | 3 - 4 | 900 | 126 | |
| BT025865.1 | - | 3 - 4 | 811 | 126 | ||
Set Membership: |
|