Search
Search

MYB86| Gene Symbol: MYB86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180548.4 | NP_850879.1 | 1 - 2 | 373 | 153 | |
| GenBank mRNA | AY099777.1 | - | 1 - 2 | 259 | 153 | |
| AY519629.1 | - | 1 - 2 | 133 | 153 | ||
| BT008740.1 | - | 1 - 2 | 133 | 153 | ||
Set Membership: |
|