Search
Search

CPK13| Gene Symbol: CPK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339535.1 | NP_001326349.1 | 4 - 5 | 1344 | 152 | |
| NM_001339536.1 | NP_001326348.1 | 4 - 5 | 1275 | 152 | ||
| NM_115044.5 | NP_190753.2 | 4 - 5 | 1344 | 152 | ||
Set Membership: |
|