Search
Search

SAR1| Gene Symbol: SAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198200.2 | NP_001185129.1 | 25 - 26 | 4502 | 93 | |
| NM_001333042.1 | NP_001319133.1 | 25 - 26 | 4490 | 93 | ||
Set Membership: |
|