Search
Search

MKK4| Gene Symbol: MKK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104044.3 | NP_175577.1 | 1 | 1447 | 147 | |
| GenBank mRNA | AB015315.1 | - | 1 | 1447 | 147 | |
| AF324667.2 | - | 1 | 1323 | 147 | ||
| AF326878.1 | - | 1 | 1323 | 147 | ||
| AF375398.1 | - | 1 | 1323 | 147 | ||
Set Membership: |
|