Search
Search

SKU5| Gene Symbol: SKU5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203775.2 | NP_001190704.1 | 7 - 8 | 1728 | 120 | |
| NM_117312.4 | NP_192979.1 | 7 - 8 | 1710 | 120 | ||
| GenBank mRNA | AF439406.1 | - | 7 - 8 | 1461 | 120 | |
Set Membership: |
|