Search
Search

EYA| Gene Symbol: EYA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129084.3 | NP_565803.1 | 5 - 6 | 800 | 72 | |
| GenBank mRNA | AF410332.1 | - | 5 - 6 | 729 | 72 | |
| AY054555.1 | - | 5 - 6 | 754 | 72 | ||
| AY064618.1 | - | 4 - 5 | 668 | 72 | ||
| AY149930.1 | - | 4 - 5 | 668 | 72 | ||
Set Membership: |
|