Search
Search

scpl5| Gene Symbol: scpl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334580.1 | NP_001319372.1 | 10 - 11 | 1081 | 148 | |
| NM_001334581.1 | NP_001323094.1 | 8 - 9 | 1247 | 148 | ||
| NM_001334582.1 | NP_001323093.1 | 10 - 11 | 1080 | 148 | ||
Set Membership: |
|