Search
Search

ALDH22A1| Gene Symbol: ALDH22A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111545.3 | NP_187321.2 | 3 - 4 | 380 | 127 | |
| NM_202513.3 | NP_974242.1 | 3 - 4 | 493 | 127 | ||
| GenBank mRNA | AJ584646.1 | - | 3 - 4 | 263 | 127 | |
Set Membership: |
|