Search
Search

SIP3| Gene Symbol: SIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119244.3 | NP_194825.1 | 1 | 1770 | 140 | |
| GenBank mRNA | AF285106.1 | - | 1 | 140 | ||
| AF339145.1 | - | 1 | 1398 | 140 | ||
| AF436831.1 | - | 1 | 1741 | 140 | ||
Set Membership: |
|