Search
Search

PBA1| Gene Symbol: PBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036682.1 | NP_001031759.1 | 5 - 6 | 586 | 67 | |
| NM_001203950.1 | NP_001190879.1 | 6 - 7 | 475 | 67 | ||
| NM_119279.5 | NP_194858.1 | 5 - 6 | 602 | 67 | ||
| GenBank mRNA | AF043529.1 | - | 5 - 6 | 571 | 67 | |
| AY052354.1 | - | 5 - 6 | 583 | 67 | ||
| AY058240.1 | - | 5 - 6 | 419 | 67 | ||
| Y13694.1 | - | 490 | 67 | |||
Set Membership: |
|