Search
Search

SYD| Gene Symbol: SYD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084502.2 | NP_001077971.1 | 5 - 6 | 786 | 142 | |
| NM_001336158.1 | NP_001325347.1 | 4 - 5 | 672 | 142 | ||
| NM_001336159.1 | NP_001325348.1 | 4 - 5 | 672 | 142 | ||
| NM_001336160.1 | NP_001325349.1 | 4 - 5 | 672 | 142 | ||
| NM_179785.3 | NP_850116.1 | 5 - 6 | 786 | 142 | ||
| NM_179786.2 | NP_850117.1 | 5 - 6 | 786 | 142 | ||
Set Membership: |
|