Search
Search

MLO4| Gene Symbol: MLO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331947.1 | NP_001320915.1 | 1 - 2 | 261 | 105 | |
| NM_001331948.1 | NP_001320916.1 | 2 - 3 | 253 | 105 | ||
| NM_100975.3 | NP_563882.1 | 1 - 2 | 269 | 105 | ||
| GenBank mRNA | AF369565.1 | - | 1 - 2 | 176 | 105 | |
Set Membership: |
|