Search
Search

FUT12| Gene Symbol: FUT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103858.5 | NP_175393.1 | 1 - 2 | 647 | 140 | |
| GenBank mRNA | AJ345085.1 | - | 1 - 2 | 386 | 140 | |
| AY054522.1 | - | 1 - 2 | 545 | 140 | ||
| BT002570.1 | - | 1 - 2 | 386 | 140 | ||
| KJ138969.1 | - | 1 - 2 | 386 | 140 | ||
Set Membership: |
|