Search
Search

MTK| Gene Symbol: MTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103869.4 | NP_564555.1 | 5 - 6 | 1219 | 97 | |
| GenBank mRNA | AY057512.1 | - | 5 - 6 | 1169 | 97 | |
| AY090315.1 | - | 5 - 6 | 1096 | 97 | ||
Set Membership: |
|