Search
Search

ABCC8| Gene Symbol: ABCC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203015.2 | NP_001189944.1 | 2 - 3 | 2356 | 106 | |
| NM_001338517.1 | NP_001326627.1 | 3 - 4 | 2267 | 106 | ||
| NM_001338518.1 | NP_001326629.1 | 2 - 3 | 2246 | 106 | ||
| NM_001338519.1 | NP_001326630.1 | 2 - 3 | 2127 | 106 | ||
| NM_001338520.1 | NP_001326628.1 | 3 - 4 | 2038 | 106 | ||
| NM_113020.3 | NP_188762.3 | 2 - 3 | 2237 | 106 | ||
Set Membership: |
|