Search
Search

CYCP2;1| Gene Symbol: CYCP2;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113083.5 | NP_188825.1 | 1 - 2 | 535 | 68 | |
| GenBank mRNA | BT014779.1 | - | 1 - 2 | 396 | 68 | |
| BT015746.1 | - | 1 - 2 | 390 | 68 | ||
Set Membership: |
|