Search
Search

BCAT3| Gene Symbol: BCAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125330.1 | NP_001118802.1 | 6 - 7 | 905 | 73 | |
| NM_114828.6 | NP_566923.1 | 6 - 7 | 1018 | 73 | ||
| GenBank mRNA | AF446355.1 | - | 6 - 7 | 883 | 73 | |
| AJ276124.1 | - | 6 - 7 | 885 | 73 | ||
| AY097417.1 | - | 6 - 7 | 792 | 73 | ||
Set Membership: |
|