Search
Search

AGL26| Gene Symbol: AGL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343985.1 | NP_001318659.1 | 4 - 5 | 487 | 136 | |
| GenBank mRNA | AK226438.1 | - | 4 - 5 | 467 | 136 | |
| AK227520.1 | - | 4 - 5 | 475 | 136 | ||
| BT010906.1 | - | 4 - 5 | 456 | 136 | ||
Set Membership: |
|