Search
Search

ABCB27| Gene Symbol: ABCB27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344283.1 | NP_001332027.1 | 4 - 5 | 405 | 98 | |
| NM_123266.8 | NP_198720.2 | 4 - 5 | 469 | 98 | ||
| GenBank mRNA | AK229810.1 | - | 4 - 5 | 351 | 98 | |
| DQ469798.1 | - | 4 - 5 | 269 | 98 | ||
Set Membership: |
|