Search
Search

HRB1| Gene Symbol: HRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344827.1 | NP_001332206.1 | 2 - 3 | 422 | 151 | |
| NM_124300.4 | NP_199734.2 | 2 - 3 | 472 | 151 | ||
| GenBank mRNA | AF372963.1 | - | 2 - 3 | 271 | 151 | |
| AY133584.1 | - | 2 - 3 | 213 | 151 | ||
| HQ322385.1 | - | 2 - 3 | 213 | 151 | ||
Set Membership: |
|