Search
Search

COI1| Gene Symbol: COI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129552.4 | NP_565919.1 | 1 - 2 | 868 | 151 | |
| GenBank mRNA | AF036340.1 | - | 1 - 2 | 752 | 151 | |
| AY045625.1 | - | 1 - 2 | 690 | 151 | ||
| AY133556.1 | - | 1 - 2 | 490 | 151 | ||
Set Membership: |
|