Search
Search

EBNA1BP2| Gene Symbol: EBNA1BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034417.2 | NP_001029589.1 | 7 - 8 | 879 | 102 | |
| GenBank mRNA | BC102356.1 | - | 7 - 8 | 880 | 102 | |
Set Membership: |
|