Search
Search

RNASEH2A| Gene Symbol: RNASEH2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038534.2 | NP_001033623.1 | 8 - 9 | 1312 | 77 | |
| GenBank mRNA | BC109682.1 | - | 8 - 9 | 1313 | 77 | |
Set Membership: |
|