Search
Search

RAMP2| Gene Symbol: RAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098860.3 | NP_001092330.1 | 2 - 3 | 280 | 76 | |
| GenBank mRNA | BC146273.1 | - | 2 - 3 | 281 | 76 | |
| DT719941.1 | - | 2 - 3 | 258 | 76 | ||
| EE245593.1 | - | 2 - 3 | 201 | 76 | ||
Set Membership: |
|