Search
Search

RDH5| Gene Symbol: RDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174166.3 | NP_776591.2 | 2 - 3 | 417 | 65 | |
| GenBank mRNA | EV670727.1 | - | 2 - 3 | 419 | 65 | |
| L36533.1 | - | 2 - 3 | 418 | 65 | ||
| X82262.1 | - | 346 | 65 | |||
Set Membership: |
|