Search
Search

HDGFL3| Gene Symbol: HDGFL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191368.2 | NP_001178297.1 | 3 - 4 | 647 | 108 | |
| GenBank mRNA | BC151802.1 | - | 3 - 4 | 722 | 108 | |
| EE216797.1 | - | 4 - 5 | 108 | |||
| EH202588.1 | - | 2 - 3 | 182 | 108 | ||
Set Membership: |
|