Search
Search

lgc-35| Gene Symbol: lgc-35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027267.5 | NP_001022438.1 | 9 - 10 | 1264 | 104 | |
| NM_001373808.1 | NP_001359643.1 | 10 - 11 | 1437 | 104 | ||
Set Membership: |
|