Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Ce02419366_m1
          See other SLO-1 GE Assays ›
          Gene Symbol:
          slo-1
          Gene Name:
          protein-coding
          Gene Aliases:
          CELE_Y51A2D.19
          Chromosome Location:
          Chr.V: 18492261 - 18508212 on Build WBcel235
          Species:
          Caenorhabditis elegans
          Species Specific ID (Flybase ID):
          -
          Assay ID Ce02419366_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: slo-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029088.2 NP_001024259.1 15 - 16 2628 70
          NM_001029089.3 NP_001024260.1 14 - 15 2568 70
          NM_001029090.5 NP_001024261.1 15 - 16 2607 70
          NM_001269872.3 NP_001256801.1 16 - 17 2607 70
          NM_001269873.3 NP_001256802.1 16 - 17 2649 70
          NM_001269874.3 NP_001256803.1 15 - 16 2583 70
          NM_001269875.4 NP_001256804.1 16 - 17 2673 70
          NM_001269876.3 NP_001256805.1 15 - 16 2589 70
          NM_001269877.3 NP_001256806.1 14 - 15 2523 70
          NM_001269878.3 NP_001256807.1 16 - 17 2649 70
          NM_001269879.4 NP_001256808.1 15 - 16 2628 70
          NM_001269880.3 NP_001256809.1 16 - 17 2628 70
          NM_001269881.3 NP_001256810.1 15 - 16 2562 70
          GenBank mRNA AF431891.1 - 15 - 16 2589 70
          AF431892.1 - 14 - 15 2523 70
          AF431893.1 - 15 - 16 2562 70
          JQ582463.1 - 16 - 17 2649 70
          JQ606809.1 - 16 - 17 2628 70
          JQ606810.1 - 14 - 15 2523 70
          JQ606812.1 - 15 - 16 2562 70

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          potassium ion transport
          ion transmembrane transport
          regulation of ion transmembrane transport
          locomotion
          response to drug
          pharyngeal pumping
          sarcomere organization
          regulation of neurotransmitter secretion
          behavioral response to ethanol
          modulation of synaptic transmission
          transmembrane transport
          potassium ion transmembrane transport
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated potassium channel activity
          potassium channel activity
          protein binding
          calcium-activated potassium channel activity
          large conductance calcium-activated potassium channel activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline